Z-Glu-Gly-OH

Supplier: BACHEM AMERICAS INC

4003632.0001 4003632.0005
C-1640.0001BAEA 317.74 CAD
C-1640.0001BA C-1640.0005BA
Z-Glu-Gly-OH
Z-Glu-Gly-OH

Sequence: Z-Glu-Gly-OH

Formula: C₁₅H₁₈N₂O₇
Storage Temperature: Freezer
MDL Number: MFCD00191093
CAS Number: 1634-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR