Z-Gln-Gly-OH ≥99% (by TLC)

Supplier: BACHEM AMERICAS INC

Synonyms: N-Benzyloxycarbonyl-L-glutaminylglycine, (S)-2-(5-Amino-2-(((benzyloxy)carbonyl)amino)-5-oxopentanamido)acetic acid

4002246.0001 4002246.0005
C-1635.0001BAEA 317.74 CAD
C-1635.0001BA C-1635.0005BA
Z-Gln-Gly-OH ≥99% (by TLC)
Z-Gln-Gly-OH

Substrate for tissue transaminase.


Direct continuous spectrophotometric assay for transglutaminase (TGase) activity.


Substrate for tissue transaminase. De Macédo et al. described a direct continuous spectrophotometric assay for transglutaminase (TGase) activity.

Formula: C₁₅H₁₉N₃O₆
Melting Pt: 187 °C
Storage Temperature: Freezer
MDL Number: MFCD00055926
CAS Number: 6610-42-0

Order Now

Specification Test Results

Identification (TLC) Complies with Authentic Material
Purity (TLC) 99%
Identification (Elemental Analysis), C 53.4%
Identification (Elemental Analysis), H 5.7%
Identification (Elemental Analysis), N 12.5%
Assay (Elemental Analysis) 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR