N-Carbobenzoxy-L-glutamine
Supplier: BACHEM AMERICAS INC
Synonyms:
N-Cbz-L-glutamine, Z-Gln-OH
4001309.0025
4001309.0100
C-1615.0025BAEA
175.66
CAD
C-1615.0025BA
C-1615.0100BA
N-Carbobenzoxy-L-glutamine
N-Carbobenzoxy-L-glutamine
Sequence: Z-Gln-OH
For isoglutamine derivatives please see the family "Glutamic Acid".
Substrate for Bacillus circulans peptidoglutaminase.
Formula:
C₁₃H₁₆N₂O₅ MW: 280.28 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00008043 CAS Number: 2650-64-8 EINECS: 220-173-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...