Z-Glu(OtBu)-OSu

Supplier: BACHEM AMERICAS INC

4000847.0005 4000847.0025
C-1605.0005BAEA 252.28 CAD
C-1605.0005BA C-1605.0025BA
Z-Glu(OtBu)-OSu
Z-Glu(OtBu)-OSu

Sequence: Z-Glu(OtBu)-OSu


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₂₁H₂₆N₂O₈
Storage Temperature: Freezer
MDL Number: MFCD00153329
CAS Number: 4666-16-4
EINECS: 225-111-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR