5-tert-Butyl-N-carbobenzoxy-L-glutamate

Supplier: BACHEM AMERICAS INC

4000475.0005 4000475.0025 4000475.0100
C-1595.0005BAEA 175.66 CAD
C-1595.0005BA C-1595.0025BA C-1595.0100BA
5-tert-Butyl-N-carbobenzoxy-L-glutamate
5-tert-Butyl-N-carbobenzoxy-L-glutamate

Sequence: Z-Glu(OtBu)-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₇H₂₃NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00038274
CAS Number: 3886-08-6
EINECS: 223-421-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR