1-Benzyl-N-carbobenzoxy-L-glutamate

Supplier: BACHEM AMERICAS INC

Synonyms: N-Cbz-L-glutamic Acid 1-Benzyl Ester, N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, Z-Glu-OBzl, 1-Benzyl N-Cbz-L-glutamate, N-Carbobenzoxy-L-glutamic Acid 1-Benzyl Ester

4000670.0005 4000670.0025
C-1570.0005BAEA 238.26 CAD
C-1570.0005BA C-1570.0025BA
1-Benzyl-N-carbobenzoxy-L-glutamate
1-Benzyl-N-carbobenzoxy-L-glutamate

Sequence: Z-Glu-OBzl


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


Educt for the γ-modification of glutamic acid, e.g. for obtaining the 1- and 2-adamantyl ester, acid-labile γ-carboxyl protecting groups. Feichtinger et al. obtained δ,ω,ω'-protected arginine from Z-Glu-OBzl via reduction to the alcohol followed by guanidinylation under Mitsunobu conditions. Additionally, Z-Glu-OBzl has been used as highly effective acyl donor in enzymatic peptide synthesis.

Formula: C₂₀H₂₁NO₆
MW: 371.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077013
CAS Number: 3705-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR