(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid
Supplier: BACHEM AMERICAS INC
Synonyms:
Cbz-Glu-OH, Z-Glu-OH, N-Cbz-L-glutamic Acid
4000114.0025
4000114.0100
C-1560.0025BAEA
175.66
CAD
C-1560.0025BA
C-1560.0100BA
(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid
(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid
Sequence: Z-Glu-OH
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
Formula:
C₁₃H₁₅NO₆ MW: 281.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002801 CAS Number: 1155-62-0 EINECS: 214-584-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...