Z-Dehydro-Ala-OMe

Supplier: BACHEM AMERICAS INC

Synonyms: Methyl 2-(((benzyloxy)carbonyl)amino)acrylate, Z-Dehydroalanine methyl ester

4008360.0001 4008360.0005
C-1540.0001BAEA 387.21 CAD
C-1540.0001BA C-1540.0005BA
Z-Dehydro-Ala-OMe
Z-Dehydro-Ala-OMe

Sequence: Z-dehydro-Ala-OMe


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).


Synonym(s): Z-Dha-OMe

Formula: C₁₂H₁₃NO₄
MW: 235.239 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00056245
CAS Number: 21149-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR