AmbotzZAA1249

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Thr(tBu)-OSu

4001247.0001 4001247.0005 4001247.0025
C-1490.0001BAEA 175.66 CAD
C-1490.0001BA C-1490.0005BA C-1490.0025BA
AmbotzZAA1249
AmbotzZAA1249

Sequence: Z-Thr(tBu)-OSu


Bachem also offers phosphothreonine derivatives.

Formula: C₂₀H₂₆N₂O₇
Storage Temperature: Freezer
MDL Number: MFCD00153346
CAS Number: 10068-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR