Z-Thr(tBu)-OH·DCHA
Supplier: BACHEM AMERICAS INC
Synonyms:
N-Benzyloxycarbonyl-O-tert-butyl-L-threonine dicyclohexylammonium salt, Z-Thr(tBu)-OH·DCHA, Dicyclohexylamine (2S,3R)-2-(((benzyloxy)carbonyl)amino)-3-(tert-butoxy)butanoate
4001246.0005
4001246.0025
4001246.0100
C-1475.0005BAEA
227.28
CAD
C-1475.0005BA
C-1475.0025BA
C-1475.0100BA
Z-Thr(tBu)-OH·DCHA
Z-Thr(tBu)-OH·DCHA
Sequence: Z-Thr(tBu)-OH · DCHA
Bachem also offers phosphothreonine derivatives.
Formula:
C₂₈H₄₆N₂O₅ MW: 490.685 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00077043 CAS Number: 16966-07-7 EINECS: 241-040-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...