Z-D-Ser(tBu)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
Cbz-D-Ser(TBu)-OH, Z-D-Ser(TBu)-OH, Nα-Benzyloxycarbonyl-O-tert-butyl-D-serine
4004207.0005
4004207.0025
C-1460.0005BAEA
601.51
CAD
C-1460.0005BA
C-1460.0025BA
Z-D-Ser(tBu)-OH
Z-D-Ser(tBu)-OH
Sequence: Z-D-Ser(tBu)-OH
Bachem also offers derivatives of phosphoserine and sulfoserine.
Formula:
C₁₅H₂₁NO₅ MW: 295.335 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00077038 CAS Number: 65806-90-8 EINECS: 480-690-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...