N-Carbobenzoxy-O-tert-butyl-L-serine

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Ser(tBu)-OH, N-Cbz-O-tert-butyl-L-serine, (S)-2-(((Benzyloxy)carbonyl)amino)-3-(tert-butoxy)propanoic acid, Cbz-Ser-OtBu

4001692.0005 4001692.0025
C-1455.0005BAEA 175.66 CAD
C-1455.0005BA C-1455.0025BA
N-Carbobenzoxy-O-tert-butyl-L-serine
N-Carbobenzoxy-O-tert-butyl-L-serine

Sequence: Z-Ser(tBu)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₁₅H₂₁NO₅
MW: 295.34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038275
CAS Number: 1676-75-1
EINECS: 216-827-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR