N-Cbz-N'-boc-L-ornithine

Supplier: BACHEM AMERICAS INC

4000362.0005 4000362.0025
C-1450.0005BAEA 291.08 CAD
C-1450.0005BA C-1450.0025BA
N-Cbz-N'-boc-L-ornithine
N-Cbz-N'-boc-L-ornithine

Sequence: Z-Orn(Boc)-OH

Formula: C₁₈H₂₆N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00038268
CAS Number: 7733-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR