Z-Lys(Boc)-ONp

Supplier: BACHEM AMERICAS INC

Synonyms: Nα-Benzyloxycarbonyl-Nepsilon-Boc-L-lysine 4-nitrophenyl ester

4001873.0005 4001873.0025
C-1440.0005BAEA 299.43 CAD
C-1440.0005BA C-1440.0025BA
Z-Lys(Boc)-ONp
Z-Lys(Boc)-ONp

Sequence: Z-Lys(Boc)-ONp


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₅H₃₁N₃O₈
MW: 501.536 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00076963
CAS Number: 2212-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR