Z-Lys(Boc)-OSu

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2,5-Dioxopyrrolidin-1-yl 2-(((benzyloxy)carbonyl)amino)-6-((tert-butoxycarbonyl)amino)hexanoate

4001830.0005 4001830.0025
C-1435.0005BAEA 299.43 CAD
C-1435.0005BA C-1435.0025BA
Z-Lys(Boc)-OSu
Z-Lys(Boc)-OSu

Sequence: Z-Lys(Boc)-OSu


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₃H₃₁N₃O₈
MW: 477.514 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038822
CAS Number: 3338-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR