Z-Lys(Boc)-OH·DCHA

Supplier: BACHEM AMERICAS INC

Synonyms: Cbz-Lys(Boc)-OH·DCHA, Z-Lys(Boc)-OH·DCHA, Z-Lys(Boc)-OH.DCHA

4001617.0005 4001617.0025
C-1425.0005BAEA 180.07 CAD
C-1425.0005BA C-1425.0025BA
Z-Lys(Boc)-OH·DCHA
Z-Lys(Boc)-OH·DCHA

Sequence: Z-Lys(Boc)-OH · DCHA


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₃₁H₅₁N₃O₆
MW: 561.764 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038269
CAS Number: 2212-76-2
EINECS: 218-663-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR