Nε-(tert-Butoxycarbonyl)-Nɑ-carbobenzoxy-L-lysine

Supplier: BACHEM AMERICAS INC

Synonyms: Nε-Boc-Nɑ-Cbz-L-lysine, Z-Lys(Boc)-OH

4001459.0005 4001459.0025 4001459.0100
C-1420.0005BAEA 206.6 CAD
C-1420.0005BA C-1420.0025BA C-1420.0100BA
Nε-(tert-Butoxycarbonyl)-Nɑ-carbobenzoxy-L-lysine
Nepsilon-(tert-Butoxycarbonyl)-Nalpha-carbobenzoxy-L-lysine

Sequence: Z-Lys(Boc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₉H₂₈N₂O₆
MW: 380.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00062271
CAS Number: 2389-60-8
EINECS: 219-223-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR