Cbz-Ser(Bzl)-OH

Supplier: BACHEM AMERICAS INC

4003805.0005 4003805.0025
C-1400.0005BAEA 283.5 CAD
C-1400.0005BA C-1400.0025BA
Cbz-Ser(Bzl)-OH
Cbz-Ser(Bzl)-OH

Sequence: Z-Ser(Bzl)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₁₈H₁₉NO₅
Storage Temperature: Ambient
MDL Number: MFCD00022029
CAS Number: 20806-43-3
EINECS: 244-049-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR