Z-D-Asp(OtBu)-OH hydrate

Supplier: BACHEM AMERICAS INC

4004766.0001 4004766.0005 4004766.0025
C-1360.0001BAEA 175.66 CAD
C-1360.0001BA C-1360.0005BA C-1360.0025BA
Z-D-Asp(OtBu)-OH hydrate
Z-D-Asp(OtBu)-OH hydrate

Sequence: Z-D-Asp(OtBu)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₁₆H₂₁NO₆·H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00237335
CAS Number: 71449-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR