Z-Asp(OtBu)-OH

Supplier: BACHEM AMERICAS INC

4000483.0005 4000483.0025 4000483.0100
C-1355.0005BAEA 201.47 CAD
C-1355.0005BA C-1355.0025BA C-1355.0100BA
Z-Asp(OtBu)-OH
Z-Asp(OtBu)-OH

Sequence: Z-Asp(OtBu)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₁₆H₂₁NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00042857
CAS Number: 5545-52-8
EINECS: 226-912-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR