4-Benzyl-N-carbobenzoxy-L-aspartate

Supplier: BACHEM AMERICAS INC

4001085.0005 4001085.0025 4001085.0100
C-1350.0005BAEA 175.66 CAD
C-1350.0005BA C-1350.0025BA C-1350.0100BA
4-Benzyl-N-carbobenzoxy-L-aspartate
4-Benzyl-N-carbobenzoxy-L-aspartate

Sequence: Z-Asp(OBzl)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₁₉H₁₉NO₆
Storage Temperature: Ambient
MDL Number: MFCD00037820
CAS Number: 3479-47-8
EINECS: 222-456-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR