Z-Asp-OBzl

Supplier: BACHEM AMERICAS INC

Synonyms: N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester, Cbz-Asp-OBzl

4000429.0005 4000429.0025
C-1340.0005BAEA 206.51 CAD
C-1340.0005BA C-1340.0025BA
Z-Asp-OBzl
Z-Asp-OBzl

Sequence: Z-Asp-OBzl


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.


Educt for the synthesis of homoserine derivatives.

Formula: C₁₉H₁₉NO₆
Storage Temperature: Ambient
MDL Number: MFCD00038594
CAS Number: 4779-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR