Nɑ-Carbobenzoxy-D-arginine

Supplier: BACHEM AMERICAS INC

4009614.0005 4009614.0025
C-1295.0005BAEA 201.47 CAD
C-1295.0005BA C-1295.0025BA
Nɑ-Carbobenzoxy-D-arginine
Nalpha-Carbobenzoxy-D-arginine

Sequence: Z-D-Arg-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₁₄H₂₀N₄O₄
Storage Temperature: Ambient
MDL Number: MFCD00063009
CAS Number: 6382-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR