Z-Ala-Ala-OH

Supplier: BACHEM AMERICAS INC

4001947.0005 4001947.0025
C-1045.0005BAEA 206.51 CAD
C-1045.0005BA C-1045.0025BA
Z-Ala-Ala-OH
Z-Ala-Ala-OH

Sequence: Z-Ala-Ala-OH

Formula: C₁₄H₁₈N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD00037234
CAS Number: 16012-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR