Z-Ala-OSu

Supplier: BACHEM AMERICAS INC

Synonyms: 2,5-Dioxopyrrolidin-1-yl ((benzyloxy)carbonyl)-L-alaninate

4000224.0025 4000224.0100
C-1025.0025BAEA 336.59 CAD
C-1025.0025BA C-1025.0100BA
Z-Ala-OSu
Z-Ala-OSu

Sequence: Z-Ala-OSu


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₁₅H₁₆N₂O₆
MW: 320.301 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00053548
CAS Number: 3401-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR