N-Carbobenzoxy-β-alanine
Supplier: BACHEM AMERICAS INC
Synonyms:
Carbobenzyloxy-beta-alanine
4000236.0025
4000236.0100
C-1015.0025BAEA
180.91
CAD
C-1015.0025BA
C-1015.0100BA
N-Carbobenzoxy-β-alanine
N-Carbobenzoxy-beta-alanine
Sequence: Z-β-Ala-OH
Bachem also offers Fmoc-β-Ala-Wang resin (D-1625) and Boc-β-Ala-PAM resin (D-1650).
Formula:
C₁₁H₁₃NO₄ MW: 223.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037292 CAS Number: 2304-94-1 EINECS: 218-967-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...