N-Carbobenzyloxy-L-alanine

Supplier: BACHEM AMERICAS INC

Synonyms: N-Cbz-L-alanine, Z-Ala-OH, (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid, N-Benzyloxycarbonyl-L-alanine, ((Benzyloxy)carbonyl)-L-alanine

4000246.0025 4000246.0100
C-1000.0025BAEA 175.66 CAD
C-1000.0025BA C-1000.0100BA
N-Carbobenzyloxy-L-alanine
N-Carbobenzyloxy-L-alanine

Sequence: Z-Ala-OH


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₁₁H₁₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00002640
CAS Number: 1142-20-7
EINECS: 214-532-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR