Fmoc-α-Me-D-Leu-OH

Supplier: BACHEM AMERICAS INC

4066676.0001 4066676.0005
B-4080.0001BAEA 478.3 CAD
B-4080.0001BA B-4080.0005BA
Fmoc-α-Me-D-Leu-OH
Fmoc-α-Me-D-Leu-OH

Sequence: Fmoc-α-Me-D-Leu-OH


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).

Formula: C₂₂H₂₅NO₄
Storage Temperature: Freezer
MDL Number: MFCD12031693
CAS Number: 1231709-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR