Fmoc-D-Cys(Mtt)-OH

Supplier: BACHEM AMERICAS INC

4066063.0005 4066063.0025
B-4070.0005BAEA 2519.53 CAD
B-4070.0005BA B-4070.0025BA
Fmoc-D-Cys(Mtt)-OH
Fmoc-D-Cys(Mtt)-OH

Sequence: Fmoc-D-Cys(Mtt)-OH


Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).

Formula: C₃₈H₃₃NO₄S
Storage Temperature: Freezer
MDL Number: MFCD14155676
CAS Number: 252206-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR