Fmoc-cis-4-fluoro-Pro-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (2S,4S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-fluoropyrrolidine-2-carboxylic acid, Fmoc-cis-4-F-Pro-OH

4034298.0250 4034298.1000 4034298.5000
B-4050.0250BAEA 317.74 CAD
B-4050.0250BA B-4050.1000BA B-4050.5000BA
Fmoc-cis-4-fluoro-Pro-OH
Fmoc-cis-4-fluoro-Pro-OH

Sequence: Fmoc-cis-4-fluoro-Pro-OH


Flp derivatives.


Synonym(s): Fmoc-(2S,4S)-4-fluoro-pyrrolidine-2-carboxylic acid#Fmoc-flp-OH

Formula: C₂₀H₁₈FNO₄
Storage Temperature: Freezer
MDL Number: MFCD07781261
CAS Number: 203866-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR