Fmoc-L-cysteic acid · disodium salt
Supplier: BACHEM AMERICAS INC
Sequence: Fmoc-L-cysteic acid · disodium salt
Synonym(s): Fmoc-Cya-OH · disodium salt
Protected derivative for the introduction of this highly polar amino acid. Cya-containing peptides are obtained by oxidation of cysteine or cystine, as a post-translational modification, as a by-product during peptide synthesis, or, intentionally (by using oxidants as H₂O₂/formic acid), for quantification of these sensitive amino acids. Use of B-4030 allows to obtain Cya-peptides containing sensitive amino acids, as an additional oxidation step is not required.
Formula:
C₁₈H₁₅NNa₂O₇S MW: 435.37 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02094396 CAS Number: 320384-09-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...