Fmoc-Thz-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-L-thiazolidine-4-carboxylic acid, Fmoc-L-thioproline, (R)-3-(((9H-Fluoren-9-yl)methoxy)carbonyl)thiazolidine-4-carboxylic acid, (4R)-3-[(9H-Fluoren-9-ylmethoxy)carbonyl]-1,3-thiazolane-4-carboxylic acid

4037410.0005 4037410.0025
B-4015.0005BAEA 335.58 CAD
B-4015.0005BA B-4015.0025BA
Fmoc-Thz-OH
Fmoc-Thz-OH

Sequence: Fmoc-L-thiazolidine-4-carboxylic acid


Thiazolidine-4-carboxylic acid (Thz) can be inserted as a Pro equivalent (thiaproline) as well as a masked cysteine. Contrary to Cys derivatives, Thz derivatives can be activated and coupled with minimal racemization.


Synonym(s): Fmoc-Thz-OH#Fmoc-L-thioproline

Formula: C₁₉H₁₇NO₄S
Storage Temperature: Freezer
MDL Number: MFCD00235910
CAS Number: 133054-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR