Fmoc-L-propargylglycine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Pra-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-propargylglycine, N-Fmoc-L-propargylglycine, (S)-2-(Fmoc-amino)-4-pentynoic acid, Fmoc-propargyl-Gly-OH

4053914.0001 4053914.0005
B-4000.0001BAEA 590.16 CAD
B-4000.0001BA B-4000.0005BA
Fmoc-L-propargylglycine
Fmoc-L-propargylglycine

Sequence: Fmoc-Pra-OH
Synonym(s): Fmoc-Propargyl-Gly-OH#(S)-2-(Fmoc-amino)-4-pentynoic acid


Bachem also offers resin-bound propargylglycine, Fmoc-Pra-Wang resin (D-2820).


Building block for the synthesis of precursor peptides for tritiation, and for "click chemistry". Pra-containing peptides have been modified by 1,3-dipolar cycloaddition, e.g. with alkyl azides. Peptides containing a Pra residue and an α-azido acid have been cyclized in the presence of CuBr/Na-ascorbate via triazole formation. Educt for the synthesis of Phe-derivatives by rhodium-catalyzed [2+2+2] cycloaddition.

Formula: C₂₀H₁₇NO₄
Storage Temperature: Freezer
MDL Number: MFCD01075095
CAS Number: 198561-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR