N-Fmoc-8-aminooctanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-8-Aoc-OH, Fmoc-8-aminooctanoic acid

4045043.0001 4045043.0005
B-3900.0001BAEA 206.51 CAD
B-3900.0001BA B-3900.0005BA
N-Fmoc-8-aminooctanoic acid
N-Fmoc-8-aminooctanoic acid

Sequence: Fmoc-8-aminooctanoic acid


Flexible spacer molecules of various length.


Synonym(s): Fmoc-8-Aoc-OH#Fmoc-ω-aminocaprylic acid

Formula: C₂₃H₂₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00235890
CAS Number: 126631-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR