Fmoc-N-Me-Tyr(tBu)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-α-Fmoc-N-α-methyl-O-tert-butyl-L-tyrosine

4042033.0001 4042033.0005
B-3890.0001BAEA 387.21 CAD
B-3890.0001BA B-3890.0005BA
Fmoc-N-Me-Tyr(tBu)-OH
Fmoc-N-Me-Tyr(tBu)-OH

Sequence: Fmoc-N-Me-Tyr(tBu)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₂₉H₃₁NO₅
MW: 473.569 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02684471
CAS Number: 133373-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR