(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl)amino)propanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Dap(ivDde)-OH

4054323.0001 4054323.0005
B-3885.0001BAEA 562.43 CAD
B-3885.0001BA B-3885.0005BA
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl)amino)propanoic acid
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl)amino)propanoic acid

Sequence: Fmoc-Dap(ivDde)-OH


Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.
Bachem also offers Fmoc-Dap(Dnp)-SASRIN resin (D-2590) for the SPPS of FRET substrates.


The ivDde protective group possesses higher orthogonality relatively to Fmoc in comparison to Dde.

Formula: C₃₁H₃₆N₂O₆
MW: 532.637 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01631659
CAS Number: 607366-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR