Fmoc-D-Trp(Me)-Oh

Supplier: BACHEM AMERICAS INC

4052724.0001 4052724.0005
B-3880.0001BAEA 619.6 CAD
B-3880.0001BA B-3880.0005BA
Fmoc-D-Trp(Me)-Oh
Fmoc-D-Trp(Me)-Oh

Sequence: Fmoc-D-Trp(Me)-OH


MeTrp, αMeTrp.

Formula: C₂₇H₂₄N₂O₄
Storage Temperature: Freezer
MDL Number: MFCD09037398
CAS Number: 168471-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR