Fmoc-Glu-2-phenylisopropyl ester

Supplier: BACHEM AMERICAS INC

4051412.0001 4051412.0005
B-3860.0001BAEA 542.13 CAD
B-3860.0001BA B-3860.0005BA
Fmoc-Glu-2-phenylisopropyl ester
Fmoc-Glu-2-phenylisopropyl ester

Sequence: Fmoc-Glu-2-phenylisopropyl ester


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


Synonym(s): Fmoc-Glu-OPp

Formula: C₂₉H₂₉NO₆
MW: 487.55 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10001339
CAS Number: 207305-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR