Fmoc-L-homoleucine

Supplier: BACHEM AMERICAS INC

4039689.0001 4039689.0005
B-3855.0001BAEA 516.34 CAD
B-3855.0001BA B-3855.0005BA
Fmoc-L-homoleucine
Fmoc-L-homoleucine

Sequence: Fmoc-Homoleu-OH


Hle derivatives.


Synonym(s): Fmoc-Hle-OH

Formula: C₂₂H₂₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00270206
CAS Number: 180414-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR