Fmoc-D-His(Bzl)-OH

Supplier: BACHEM AMERICAS INC

4043043.0001 4043043.0005
B-3835.0001BAEA 335.58 CAD
B-3835.0001BA B-3835.0005BA
Fmoc-D-His(Bzl)-OH
Fmoc-D-His(Bzl)-OH

Mix of τ-Bzl (mainly formed, CAS number: 1417638-37-9) and π-Bzl isomer.


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Mix of τ-Bzl (mainly formed, CAS number: 1417638-37-9) and π-Bzl isomer.

Formula: C₂₈H₂₅N₃O₄
Storage Temperature: Ambient
MDL Number: MFCD08458517
CAS Number: 1417638-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR