Fmoc-D-Ala-aldehyde

Supplier: BACHEM AMERICAS INC

4041718.0250 4041718.1000
B-3815.0250BAEA 645.24 CAD
B-3815.0250BA B-3815.1000BA
Fmoc-D-Ala-aldehyde
Fmoc-D-Ala-aldehyde

Sequence: Fmoc-D-Ala-aldehyde


Fmoc-D-alaninal.

Formula: C₁₈H₁₇NO₃
Storage Temperature: Freezer
MDL Number: MFCD08458574
CAS Number: 127043-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR