Fmoc-ala-aldehyde

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-(9H-Fluoren-9-yl)methyl (1-oxopropan-2-yl)carbamate

4025176.0001 4025176.0005
B-3810.0001BAEA 542.79 CAD
B-3810.0001BA B-3810.0005BA
Fmoc-ala-aldehyde
Fmoc-ala-aldehyde

Sequence: Fmoc-Ala-aldehyde


Fmoc-L-alaninal.

Formula: C₁₈H₁₇NO₃
MW: 295.338 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01318541
CAS Number: 146803-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR