Fmoc-7-Ahp-OH

Supplier: BACHEM AMERICAS INC

4042197.0005 4042197.0025
B-3765.0005BAEA 413.58 CAD
B-3765.0005BA B-3765.0025BA
Fmoc-7-Ahp-OH
Fmoc-7-Ahp-OH

Sequence: Fmoc-7-aminoheptanoic acid


Flexible spacer molecules of various length.

Formula: C₂₂H₂₅NO₄
Storage Temperature: Freezer
MDL Number: MFCD01861353
CAS Number: 127582-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR