Fmoc-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-OH

Supplier: BACHEM AMERICAS INC

4038236.0250 4038236.1000
B-3760.0250BAEA 841.57 CAD
B-3760.0250BA B-3760.1000BA
Fmoc-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-OH
Fmoc-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-OH

Sequence: Fmoc-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-OH
Synonym(s): Fmoc-Pam₂Cys-OH


Incorporation of Fmoc-Pam₂Cys-OH yields more hydrophilic products than incorporation of Pam₃Cys-OH (F-2630) due to the free amino group obtained after attachment and deprotection. Nevertheless, the H-Pam₂-peptides show comparable adjuvant activity.

Formula: C₅₃H₈₃NO₈S
Storage Temperature: Freezer
MDL Number: MFCD00153354
CAS Number: 210532-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR