N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine hydrate

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-L-serine, Fmoc-Ser-OH

4044985.0025 4044985.0100 4044985.0250
B-3745.0025BAEA 238.85 CAD
B-3745.0025BA B-3745.0100BA B-3745.0250BA
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine hydrate
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine hydrate

Sequence: Fmoc-Ser-OH · H₂O


Bachem also offers derivatives of phosphoserine and sulfoserine.


Educt for the synthesis of O-glycosylated serine building blocks. Liu et al. synthesized O-mannosylated Fmoc-Ser.

Formula: C₁₈H₁₇NO₅
Storage Temperature: Ambient
MDL Number: MFCD04113634
CAS Number: 73724-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR