Fmoc-β-(7-methoxy-coumarin-4-yl)-Ala-OH
Supplier: BACHEM AMERICAS INC
4046868.0500
4046868.1000
B-3740.0500BAEA
688.88
CAD
B-3740.0500BA
B-3740.1000BA
Fmoc-β-(7-methoxy-coumarin-4-yl)-Ala-OH
Fmoc-β-(7-methoxy-coumarin-4-yl)-Ala-OH
Sequence: Fmoc-β-(7-methoxy-coumarin-4-yl)-Ala-OH
Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines
Synonym(s): (S)-2-(Fmoc-amino)-3-(7-methoxy-2-oxo-2H-chromen-4-yl)-propionic acid#Fmoc-β-(O-methyl-umbellifer-4-yl)-Ala-OH
Formula:
C₂₈H₂₃NO₇ Storage Temperature: Freezer |
MDL Number:
MFCD08702736 CAS Number: 524698-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...