Fmoc-aspartic acid-4-methyl ester

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-L-aspartic acid 4-methyl ester, Fmoc-Asp(OMe)-OH

4046252.0005 4046252.0025
B-3735.0005BAEA 424.25 CAD
B-3735.0005BA B-3735.0025BA
Fmoc-aspartic acid-4-methyl ester
Fmoc-aspartic acid-4-methyl ester

Sequence: Fmoc-Asp(OMe)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₂₀H₁₉NO₆
MW: 369.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08458573
CAS Number: 145038-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR