Fmoc-β-(1-piperazinyl)-Ala(Boc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxycarbonyl)piperazin-1-yl)propanoic acid

4043668.0250 4043668.1000
B-3725.0250BAEA 770.16 CAD
B-3725.0250BA B-3725.1000BA
Fmoc-β-(1-piperazinyl)-Ala(Boc)-OH
Fmoc-β-(1-piperazinyl)-Ala(Boc)-OH

Sequence: Fmoc-β-(1-piperazinyl)-Ala(Boc)-OH


Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines


Synonym(s): (S)-3-(4-Boc-piperazin-1-yl)-2-(Fmoc-amino)-propionic acid

Formula: C₂₇H₃₃N₃O₆
MW: 495.58 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01860747
CAS Number: 313052-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR