(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)propanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-alanine, Fmoc-N-Me-Ala-OH, N-Fmoc-N-methyl-L-alanine

4038888.0001 4038888.0005
B-3720.0001BAEA 258.14 CAD
B-3720.0001BA B-3720.0005BA
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)propanoic acid
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)propanoic acid

Sequence: Fmoc-N-Me-Ala-OH


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₁₉H₁₉NO₄
MW: 325.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153384
CAS Number: 84000-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR