Fmoc-N-Me-Lys(boc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-6-((tert-butoxycarbonyl)amino)hexanoic acid

4038250.0250 4038250.1000
B-3685.0250BAEA 1032.67 CAD
B-3685.0250BA B-3685.1000BA
Fmoc-N-Me-Lys(boc)-OH
Fmoc-N-Me-Lys(boc)-OH

Sequence: Fmoc-N-Me-Lys(Boc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₇H₃₄N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD04973182
CAS Number: 197632-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR